Learn More
Perkin Elmer US LLC Retrieving Wire
Catalog No. 502104327
Encompass_Preferred
PerkinElmer offers spare and replacement parts for maintenance and consumable parts shipped with their Elemental Analyzers and accessories. This wire ships standard with the 2400 CHN Accessories Kit and Alpha Accessory Kit, but with this part number, labs can order this wire as a replacement or as a spare.
Catalog No. 50-210-4327 Supplier Perkin Elmer US LLC Supplier No. N2411274
PerkinElmer offers spare and replacement parts for maintenance and consumable parts shipped with their Elemental Analyzers and accessories. This wire ships standard with the 2400 CHN Accessories Kit and Alpha Accessory Kit, but with this part number, labs can order this wire as a replacement or as a spare.