Learn More
Medchemexpress LLC Retro-2 | 1201652-50-7 | 98.0% | 5 MG
SDP
Catalog No. 5000232264
Encompass_Preferred
Retro-2 is a small-molecule inhibitor of retrograde protein trafficking that disrupts endosome-trans-Golgi network transport and has demonstrated antiviral activity in cell-based assays.
  • Selective inhibitor of retrograde protein trafficking.
  • Shows activity against filovirus/ebolavirus in cell assays (EC50 12.2 μM in HeLa cells).
  • Induces autophagy and affects intracellular trafficking pathways.
  • High purity research-grade material supplied for laboratory use.
  • Available in small research quantities for assay development and mechanistic studies.
Catalog No. 50-002-32264 Supplier Medchemexpress LLC Supplier No. HY1225715MG
Retro-2 is a small-molecule inhibitor of retrograde protein trafficking that disrupts endosome-trans-Golgi network transport and has demonstrated antiviral activity in cell-based assays.
  • Selective inhibitor of retrograde protein trafficking.
  • Shows activity against filovirus/ebolavirus in cell assays (EC50 12.2 μM in HeLa cells).
  • Induces autophagy and affects intracellular trafficking pathways.
  • High purity research-grade material supplied for laboratory use.
  • Available in small research quantities for assay development and mechanistic studies.