Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retro-2 | 1201652-50-7 | 98.0% | 50 MG
SDP

Catalog No. 5000232583
Encompass_Preferred

Retro-2 is a small-molecule inhibitor that selectively blocks retrograde protein trafficking at the endosome-trans-Golgi network interface and shows reported antiviral activity in cellular studies.

  • Selective inhibitor of retrograde protein trafficking.
  • Reported antiviral activity in cell-based assays.
  • Induces autophagy in treated cells.
  • High purity (98.0%).
  • Solid form; recommended storage at 4°C and protection from light.
  • Available in a 50 mg pack size for research use.

Catalog No. 50-002-32583 Supplier Medchemexpress LLC Supplier No. HY12257150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Retro-2 is a small-molecule inhibitor that selectively blocks retrograde protein trafficking at the endosome-trans-Golgi network interface and shows reported antiviral activity in cellular studies.

  • Selective inhibitor of retrograde protein trafficking.
  • Reported antiviral activity in cell-based assays.
  • Induces autophagy in treated cells.
  • High purity (98.0%).
  • Solid form; recommended storage at 4°C and protection from light.
  • Available in a 50 mg pack size for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.