Learn More
Medchemexpress LLC HY-114698 5mg Medchemexpress, Retro-2 cycl CAS:1429192-00-6 Purity:>98%
SDP
Catalog No. 501932497
Encompass_Preferred
Medchemexpress, HY-114698 5mg Retro-2 cycl CAS:1429192-00-6 Retro-2 cycl (RN 1-001) is a dihydroquinazolinone (DHQZ) inhibitor of retrograde trafficking. Retro-2 cycl (RN 1-001) inhibits JCPyV and HPV16 pseudovirus with IC 50 s of 54 μM and 160 μM, respectively [1] . Antiviral agent [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2497 Supplier Medchemexpress LLC Supplier No. HY1146985MG
Medchemexpress, HY-114698 5mg Retro-2 cycl CAS:1429192-00-6 Retro-2 cycl (RN 1-001) is a dihydroquinazolinone (DHQZ) inhibitor of retrograde trafficking. Retro-2 cycl (RN 1-001) inhibits JCPyV and HPV16 pseudovirus with IC 50 s of 54 μM and 160 μM, respectively [1] . Antiviral agent [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.