Learn More
Takara Bio Retro-X™ Concentrator, 100 ml
SDP
Catalog No. NC9923886
Encompass
The Retro-X Concentrator is a complete reagent for the concentration of infectious retroviral vector particles. This reagent provides a scalable alternative to ultracentrifugation for viral particle concentration. Vector supernatants can be concentrated 10–100 fold, depending upon the volumes and envelope used. 100ml
Catalog No. NC9923886 Supplier Takara Bio Supplier No. 631455
The Retro-X Concentrator is a complete reagent for the concentration of infectious retroviral vector particles. This reagent provides a scalable alternative to ultracentrifugation for viral particle concentration. Vector supernatants can be concentrated 10–100 fold, depending upon the volumes and envelope used. 100ml