Learn More
System Biosciences LLC Retro-Concentin™ Retroviral Concentration ReagentRetro-Concentin = 100 mL
SDP
Catalog No. NC1627832
Encompass_Preferred
Easily concentrate any retrovirus with Retro-Concentin, a reagent which enables ultracentrifugation-free retrovirus concentration for higher infectivity. Quantitatively precipitate any retrovirus with Retro-Concentin™. Instead of using lengthy spins in an ultracentrifuge, Retro-Concentin enables concentration of retroviruses directly from culture medium with a simple one (or an optional two) low-speed centrifugation steps. In addition, Retro-Concentin stabilizes the retroviruses for frozen storage, providing an additional advantage over other concentration methods.
Catalog No. NC1627832 Supplier System Biosciences LLC Supplier No. RV100A1
Easily concentrate any retrovirus with Retro-Concentin, a reagent which enables ultracentrifugation-free retrovirus concentration for higher infectivity. Quantitatively precipitate any retrovirus with Retro-Concentin™. Instead of using lengthy spins in an ultracentrifuge, Retro-Concentin enables concentration of retroviruses directly from culture medium with a simple one (or an optional two) low-speed centrifugation steps. In addition, Retro-Concentin stabilizes the retroviruses for frozen storage, providing an additional advantage over other concentration methods.