Learn More
Electron Microscopy Sciences DIM Option - Retrofit, 1/EA
SDP
Supplier:  Electron Microscopy Sciences SFABASEDIM
Encompass_Preferred
The DIM retrofit option adds a combined switch and dimmer control. When you first turn the unit on it is at full power. As you rotate the switch the intensity decreases, reaching about 30% power at the minimum setting. You have finer control in the brighter portion of the adjustment range. An intensity indicator makes it easy to record and repeat preferred settings.
Catalog No. 50-209-1586