Learn More
Bellco Glass, Inc. Retrofit Kit
SDP
Catalog No. 501946120
Encompass_Preferred
This is a conversion kit for old style rocker platforms only. The old style design was a mounting plate that the platform tray bolted to. This kit will make the unit to a peg and grommet design.. MADE IN THE USA
Catalog No. 50-194-6120 Supplier Bellco Glass, Inc. Supplier No. 774031020
This is a conversion kit for old style rocker platforms only. The old style design was a mounting plate that the platform tray bolted to. This kit will make the unit to a peg and grommet design.. MADE IN THE USA