Learn More
Takara Bio RETRONECTIN DISH 10 PLATES
SDP
Catalog No. 50444033
Encompass
Dish; Precoated with RetroNectin Human Recombinant Fibronectin Fragment; Source: E. coli expressing human fibronectin fragment CH-296; For retrovirus-mediated gene transduction; 10 dishes
Catalog No. 50-444-033 Supplier Takara Bio Supplier No. T110A
Dish; Precoated with RetroNectin Human Recombinant Fibronectin Fragment; Source: E. coli expressing human fibronectin fragment CH-296; For retrovirus-mediated gene transduction; 10 dishes