Learn More
Takara Bio RetroNectin GMP grade, 2.5 mL
SDP
Catalog No. NC1753984
Encompass
Enhanced gene transfer efficiency for retroviral or lentiviral transduction of suspension cells, hematopoietic stem cells, and other hard-to-infect cell types.
Catalog No. NC1753984 Supplier Takara Bio Supplier No. T202
Enhanced gene transfer efficiency for retroviral or lentiviral transduction of suspension cells, hematopoietic stem cells, and other hard-to-infect cell types.