Learn More
SAI Infusion Technologies Micro500 500uL Blood Tube- K2EDTA, Split Septum Cap
SDP
Catalog No. NC2584007
Encompass
500ul Micro500 Blood Tube with Split Septum Cap - K2EDTA. Specialized Cap accepts a microhematocrit tube for retro-orbital blood collection or a blunt needle for sample transfer. K2EDTA for Plasma collection. SAI's Micro500 blood tubes replicate the dimensions of the discontinued BD Microtainer for 500ul blood collections. With the same slim 8 mm X 43 mm size, no adjustments for methods or instrumentation are required. 100/pack. Product is for laboratory animal research only.
Catalog No. NC2584007 Supplier SAI Infusion Technologies Supplier No. M500ESS
500ul Micro500 Blood Tube with Split Septum Cap - K2EDTA. Specialized Cap accepts a microhematocrit tube for retro-orbital blood collection or a blunt needle for sample transfer. K2EDTA for Plasma collection. SAI's Micro500 blood tubes replicate the dimensions of the discontinued BD Microtainer for 500ul blood collections. With the same slim 8 mm X 43 mm size, no adjustments for methods or instrumentation are required. 100/pack. Product is for laboratory animal research only.