Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retrorsine | 480-54-6 | 99.9% | 351.39 g/mol | C18H25NO6 | 10 MG
SDP

Catalog No. 5000289482
Encompass_Preferred

Retrorsine is a naturally occurring pyrrolizidine alkaloid used as a research reagent for studying hepatocellular injury and hepatotoxicity. It binds to DNA and inhibits hepatocyte proliferation. The compound is supplied as a white to off-white solid and is intended for laboratory research applications.

  • Research use in hepatotoxicity and liver injury models
  • Binds DNA and inhibits hepatocyte proliferation
  • High purity: 99.91%
  • Molecular weight: 351.39 g/mol; formula: C18H25NO6
  • CAS number: 480-54-6
  • Appearance: white to off-white solid
  • Storage: store solid at -20°C protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month)

Catalog No. 50-002-89482 Supplier Medchemexpress LLC Supplier No. HYN663810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Retrorsine is a naturally occurring pyrrolizidine alkaloid used as a research reagent for studying hepatocellular injury and hepatotoxicity. It binds to DNA and inhibits hepatocyte proliferation. The compound is supplied as a white to off-white solid and is intended for laboratory research applications.

  • Research use in hepatotoxicity and liver injury models
  • Binds DNA and inhibits hepatocyte proliferation
  • High purity: 99.91%
  • Molecular weight: 351.39 g/mol; formula: C18H25NO6
  • CAS number: 480-54-6
  • Appearance: white to off-white solid
  • Storage: store solid at -20°C protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.