Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix RETROtek HIV-1 ICx/CRx Kit
SDP

Catalog No. 22156771 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-771 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-771 Supplier ZeptoMetrix Supplier No. 0801096
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

ICx Acid (25mL): Reagent A, ICx Base (25mL): Reagent B, ICx Positive Control (1mL): Reagent C, ICx/CRx Negative Control (1 mL): Reagent D, CRx Control Reagent (1mL): Reagent E, CRx Neutralization Reagent (1mL)

Enhances the detection of of p24 antigen in serum or plasma

  • Contains two sets of reagents
  • The Immune Complex Dissociation (ICx) Reagents may identify serum or plasma specimens in which p24 antigen is masked by antibody
  • The Confirmation Reagents (CRx) may confirm the reactivity of positive specimens

For Immune Dissociation and Reactivity Confirmation

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Liquid
Product Type HIV Type 1 p24 ICx/CRx kit
For Use With (Application) RETRO-TEK HIV-1 p24 Antigen ELISA
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.