Learn More
101 BIO Retrovirus 10X Titer-Up Reagent (Increase virus titer by 10 times) | 1 mL for packaging 500 mL retrovirus soup
Catalog No. NC1692641
Encompass
This retrovirus titer boost is a novel recipe of small molecules designed for increasing / boosting retrovirus packaging efficiency. Retrovirus 10X Titer-Up stably regulates the viral RNA packaging on the transcriptional level, which can enhance production of retroviral particles for up to 10 fold.
Catalog No. NC1692641 Supplier 101 BIO Supplier No. P909
This retrovirus titer boost is a novel recipe of small molecules designed for increasing / boosting retrovirus packaging efficiency. Retrovirus 10X Titer-Up stably regulates the viral RNA packaging on the transcriptional level, which can enhance production of retroviral particles for up to 10 fold.