Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Reusable Barrier Lab Coats

Catalog No. 18111295
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-295 6X-Large
18-111-286 X-Small
18-111-287 Small
18-111-288 Medium
18-111-289 Large
18-111-290 X-Large
18-111-291 2X-Large
18-111-292 3X-Large
18-111-293 4X-Large
18-111-294 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 18-111-295 Supplier CritiCore Protective Wear Supplier No. 12L16403BARI016XLWHT
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Specialized lab coats provide optimal fluid resistance. High-density construction protects from permeability.

  • 100% polyester fluid-resistant front and sleeves
  • Quick-release snap front closure
  • Two front pockets and one left breast pocket
  • Side seam openings for pants pocket access
  • Splash Resistant Polyester front and sleeves
  • Breathable PolyCotton back panel for ventilation
  • Industrial launderable
  • Can be used as part of an OSHA Compliance Plan

Specifications

Product Type General Purpose Lab Coat
Size 6X-Large
Gender Unisex
Material Polycotton
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids
Style Full-length
No. of Pockets 3
Description Fluid-Resistant Barrier Front, Poly 65 Cotton 35 Back
Collar Type Mandarin
Latex Free Yes
For Use With (Application) Healthcare, Dental, Laboratory, Medical, Veterinary
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.