Learn More
CritiCore Protective Wear Reusable Barrier Lab Coats with Static Management
Catalog No. 18111299
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-299 Large
18-111-296 X-Small
18-111-297 Small
18-111-298 Medium
18-111-300 X-Large
18-111-301 2X-Large
18-111-302 3X-Large
18-111-303 4X-Large
18-111-304 5X-Large
18-111-305 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 18-111-299 Supplier CritiCore Protective Wear Supplier No. 12L25059BARE3PLWHT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to provide 360-degree splash protection, this barrier coat is made with high-density, fluid-resistant polyester fabric and carbon fiber stripes to dissipate static electricity.
  • 99% Polyester, 1% Carbon fabric
  • Splash Resistant Polyester fabric with Carbon Fiber stripes
  • Quick-release, concealed snap front closure
  • Two front pockets and one breast pocket
  • Side seam openings for pants pocket access
  • Fluid-resistant front, sleeves, and back panel
  • Knit cuffs with carbon to manage static electricity
  • Can be used as part of an OSHA Compliance Plan
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Polyester
Color White (with thin black carbon-fiber stripes)
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Static
No. of Pockets 3
Collar Type Lapel
Latex Free Yes