Learn More
World Precision Instrument Reusable biopsy punch kit
SDP
Supplier:  World Precision Instrument WP2020F
Encompass
The Rapid-Punch consists of a razor sharp cutting tip designed to cut, retrieve and store cored samples from source materials such as tissue, gels, paper, cloth, leaves, paint chips, films or other thin substrates. The Rapid-Punch is recommended as the special DNA sampling tool. It is ideal for forensic and most laboratory sampling purposes. Designed for heavy, numerous and precise sampling operations. It is durable, lasting up to 2000 punches.
Catalog No. 50-253-8333