Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Catalog No. R503155 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Inoculating Loop
Inoculating Loop Calibration Gauge
Inoculating Needle
Inoculating TB Spade
Insulated Brass Holder
Teasing Needle
Handle Type:
Blue Insulated Handle
Brass Loop Holder
Green Insulated Handle
Not Applicable
Orange Insulated Handle
Purple Insulated Handle
White Insulated Handle
Without Handle
Yellow Insulated Handle
Size:
0.001 mL
1/100 mL
1/1000 mL
2 mm
3 mm
5 mm
6 in.
8 in.
Quantity:
25/Pk.
5/Pk.
Each
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Handle Type Size Quantity
R503155 Inoculating Loop Yellow Insulated Handle 5 mm Each
R503060 Inoculating Needle Purple Insulated Handle - Each
R503152 Inoculating Loop White Insulated Handle 2 mm 5/Pk.
R503154 Inoculating Loop Green Insulated Handle 3 mm Each
R503620 Inoculating Loop Without Handle 1/1000 mL Each
R501966 Insulated Brass Holder Brass Loop Holder 6 in. Each
R501968 Insulated Brass Holder Brass Loop Holder 8 in. Each
R503070 Teasing Needle Yellow Insulated Handle - Each
R503080 Teasing Needle Orange Insulated Handle - Each
R503090 Inoculating TB Spade Blue Insulated Handle - Each
R503153 Inoculating Loop Green Insulated Handle 3 mm Each
R503156 Inoculating Loop Yellow Insulated Handle 5 mm 5/Pk.
R503513 Inoculating Loop Without Handle 3 mm 25/Pk.
R503515 Inoculating Loop Without Handle 5 mm 25/Pk.
R503630 Inoculating Loop Without Handle 1/100 mL Each
R503730 Inoculating Loop Blue Insulated Handle 1/1000 mL Each
R503740 Inoculating Loop Orange Insulated Handle 1/100 mL Each
R507020 Inoculating Loop Calibration Gauge Not Applicable 0.001 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. R503155 Supplier Thermo Scientific™ Supplier No. R503155
Add to Cart
Add to Cart
This item is not returnable. View return policy
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
  • Twisted nichrome made
  • With or without insulated handle
  • Available in different color codes
TRUSTED_SUSTAINABILITY
Specifications
Material Nichrome
Size 5 mm
Product Type Inoculating Loop
Quantity Each
Handle Type Yellow Insulated Handle