Learn More
CritiCore Protective Wear Reusable Double-Layer Integrity 1800/Tricot Face Mask (Imported)
Catalog No. 18111380
Change view
Click to view available options
Color: White
Catalog No.
18-111-380 Color: White
1 options
This item is not returnable. View return policy
Catalog No. 18-111-380 Supplier CritiCore Protective Wear Supplier No. PD 6677
This item is not returnable. View return policy
General-purpose face mask not suitable for virologic or bacterial protection.
  • Washable
  • 100% filament polyester
  • One layer of Integrity 1800 cleanroom fabric
  • One layer of Tricot surgical fabric
Specifications
Product Type General Purpose Face Mask
Color White
Fastening Type Ties
Material Tricot Fabric
Model 11FM6676N18001