Learn More
Hillrom Reusable Ear Specula
Catalog No. 22355154
Change view
Click to view available options
For Use With (Equipment):
MacroView Otoscope, and Diagnostic Otoscope
Catalog No. For Use With (Equipment)
22-355-154 MacroView Otoscope, and Diagnostic Otoscope
1 options
This item is not returnable. View return policy
Catalog No. 22-355-154 Supplier Hillrom Supplier No. WEL 24303U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Reusable Ear Specula for MacroView and Diagnostic Otoscopes
  • The specula may be autoclaved, boiled, or sterilized in solution and can be attached easily
  • For use with MacroView and Diagnostic Otoscopes
  • Length: 3 mm
  • Plastic, Round Tip
Specifications
For Use With (Equipment) MacroView Otoscope, and Diagnostic Otoscope
Size (Metric) 3 mm
Product Type Ear Specula