Learn More
MARIAN MEDICAL INC Reusable ENFit Low Dose Syringe, 5 ml, 100 Per Box
Catalog No. NC3958224
Encompass_Preferred
Enteral Syringe. Reusable Low Dose Syringe. Marian Medical Inc. MH Series. Reusable ENFit low dose syringe without offset port. Manufactured to ISO 80369 3 connection standards. Sterile individually packaged syringe. Designed for accurate small volume medication delivery. For enteral medication administration in clinical and home care settings. Packaged per box.
Catalog No. NC3958224 Supplier MARIAN MEDICAL INC Supplier No. MH05
Enteral Syringe. Reusable Low Dose Syringe. Marian Medical Inc. MH Series. Reusable ENFit low dose syringe without offset port. Manufactured to ISO 80369 3 connection standards. Sterile individually packaged syringe. Designed for accurate small volume medication delivery. For enteral medication administration in clinical and home care settings. Packaged per box.