Learn More
MARIAN MEDICAL INC Reusable ENFit Syringe, 60 ml, Offset Port, 60 Per Box
Catalog No. NC3958221
Encompass_Preferred
Enteral Syringe. Reusable Syringe. Marian Medical Inc. MH Series. Reusable ENFit syringe with offset port design. Manufactured to ISO 80369 3 connection standards. Sterile individually packaged syringe. Clear barrel with precise graduations. For enteral medication administration in clinical and home care settings. Packaged per box.
Catalog No. NC3958221 Supplier MARIAN MEDICAL INC Supplier No. MH60
Enteral Syringe. Reusable Syringe. Marian Medical Inc. MH Series. Reusable ENFit syringe with offset port design. Manufactured to ISO 80369 3 connection standards. Sterile individually packaged syringe. Clear barrel with precise graduations. For enteral medication administration in clinical and home care settings. Packaged per box.