Learn More
Medicus Health Reusable Flat Cleaning Brushes for Surgical Instruments, Extra Rigid Bristles, Blue
SDP
Catalog No. NC3309110
Encompass
Reusable Flat Cleaning Brushes for Surgical Instruments are specifically designed for hospital use in the pre-clean and preparation of surgical instruments
∙ Autoclavable
∙ Long ergonomic handle permits ease of use
∙ 3 Brushes per Pack∙ Extra Rigid Bristle Brushes (Blue): Instrument safe polyamide fibers offer steel-like rigidity and allows deep cleaning of soiled instruments, particularly in grooved or uneven surface ∙ Brush Size is 75mm x 20mm x 10mm.
∙ Brush length is 220 mm
∙ Extra long length of brush head (75 [±5] mm in Reusable)
∙ Transparent brush fibers allow for quick verification of its cleanliness
∙ Polyamide fiber bristles
∙ Plastic handle will not release any toxic components
∙ Designed to protect medical instruments from damage during sterilization, storage, and handling
Catalog No. NC3309110 Supplier Medicus Health Supplier No. 4026M4
Reusable Flat Cleaning Brushes for Surgical Instruments are specifically designed for hospital use in the pre-clean and preparation of surgical instruments
∙ Autoclavable
∙ Long ergonomic handle permits ease of use
∙ 3 Brushes per Pack∙ Extra Rigid Bristle Brushes (Blue): Instrument safe polyamide fibers offer steel-like rigidity and allows deep cleaning of soiled instruments, particularly in grooved or uneven surface ∙ Brush Size is 75mm x 20mm x 10mm.
∙ Brush length is 220 mm
∙ Extra long length of brush head (75 [±5] mm in Reusable)
∙ Transparent brush fibers allow for quick verification of its cleanliness
∙ Polyamide fiber bristles
∙ Plastic handle will not release any toxic components
∙ Designed to protect medical instruments from damage during sterilization, storage, and handling