Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC AbilityOne Reusable Flat Mop, Microfiber, 18 inch, Blue, 12 Pack

Supplier:  MSC 40128639

Encompass

This reusable flat mop is made of microfiber material and measures 18 inches. It is supplied in a pack of 12.

  • Reusable design
  • Flat mop type
  • Microfiber construction
  • 18 inch size
  • Blue color
  • Supplied in a 12-pack

Catalog No. 50-283-7351


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.