Learn More
Research Products International Corp Reusable Silica Gel Desic Bgs
SDP
Catalog No. 502388450
Encompass
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators. Durable microfiber bag securely contains beads eliminating the risk of leakage and is unaffected by moisture. Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed. Saturated gel can be regenerated by microwaving bag for 6 minutes or Regenerate at 150°C (blue color changes to pink_ May be regenerated repeatedly without degradation. Supplied individually sealed in moisture-proof pouch.
Catalog No. 50-238-8450 Supplier Research Products International Corp Supplier No. 420106A
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators. Durable microfiber bag securely contains beads eliminating the risk of leakage and is unaffected by moisture. Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed. Saturated gel can be regenerated by microwaving bag for 6 minutes or Regenerate at 150°C (blue color changes to pink_ May be regenerated repeatedly without degradation. Supplied individually sealed in moisture-proof pouch.