Learn More
MATTERHACKERS INC MatterHackers Reusable Spool Holder
SDP
Catalog No. NC3789957
Encompass
The MatterHackers ABS 3D printed, refillable spool is designed to reduce plastic waste and can accommodate up to 1kg of filament. It is compatible with Bambu refill spools and features an eye-catching hexagonal fill pattern for enhanced strength and durability. It includes a male side, a female side, and spacing shims for easy setup.Compatible with Bambu refills1kg capacityHexagonal fill pattern
Catalog No. NC3789957 Supplier MATTERHACKERS INC Supplier No. M12JVJ0H
The MatterHackers ABS 3D printed, refillable spool is designed to reduce plastic waste and can accommodate up to 1kg of filament. It is compatible with Bambu refill spools and features an eye-catching hexagonal fill pattern for enhanced strength and durability. It includes a male side, a female side, and spacing shims for easy setup.Compatible with Bambu refills1kg capacityHexagonal fill pattern