Learn More
Braintree Scientific Reusable Stainless Feeding Needle, 18G-1"""" straight 4/pk
SDP
Catalog No. 501955392
Encompass
Re-usable Animal Feeding Needles are designed to meet the needs of pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals. Precision assembly of stainless steel balls and tubing insures a quality needle suited for feeding and dosing without injury to sensitive tissues. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5392 Supplier Braintree Scientific Supplier No. NPK008
 Restricted Item
Add to Cart
Add to Cart
Re-usable Animal Feeding Needles are designed to meet the needs of pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals. Precision assembly of stainless steel balls and tubing insures a quality needle suited for feeding and dosing without injury to sensitive tissues. **Sold for research purposes only and not for human use**