Learn More
GLOMED INC Reusable Animal Feeding Needle 24G x 25mm Straight
Catalog No. NC1304166
Encompass_Preferred
Reusable Metal Hub with luer lock fitting - Smooth stainless steel ball tip and stainless steel tubing - Can be used as perfusion needle - Packed individually - Sold unsterile - please sterilize before use
Catalog No. NC1304166 Supplier GLOMED INC Supplier No. AFN2425S
Reusable Metal Hub with luer lock fitting - Smooth stainless steel ball tip and stainless steel tubing - Can be used as perfusion needle - Packed individually - Sold unsterile - please sterilize before use