Learn More
GLOMED INC Reusable Animal Feeding Needle 20G x 25mm Curved
Catalog No. NC1197790
Encompass_Preferred
Reusable Animal Feeding Needle (Crop Needle / Oral Gavage Needle) with smooth ball tip 20G tubing 25mm Length 2mm ball tip diameter Curved tubing. Please note image in listing may be slightly different than actual product depending on the size of the needle.
This item is not returnable. View return policy
Catalog No. NC1197790 Supplier GLOMED INC Supplier No. AFN2025C
 Restricted Item
Add to Cart
Add to Cart
Reusable Animal Feeding Needle (Crop Needle / Oral Gavage Needle) with smooth ball tip 20G tubing 25mm Length 2mm ball tip diameter Curved tubing. Please note image in listing may be slightly different than actual product depending on the size of the needle.