Learn More
Roboz Surgical REUSEABLE FEEDING NEEDLE 18GR
SDP
Catalog No. NC9390928
Encompass
This reusable feeding needle is designed for precise laboratory applications, featuring a curved 18-gauge stainless steel construction for durability and ease of use. It is engineered with a 2-inch length and a 2.25 mm ball diameter, providing reliable performance for animal research tasks.
  • Constructed from high-quality stainless steel for long-term reusability.
  • Features a curved design to facilitate targeted administration.
  • Equipped with a 2.25 mm ball tip for safe insertion.
  • Compatible with standard laboratory procedures requiring 18-gauge dimensions.
Catalog No. NC9390928 Supplier Roboz Surgical Supplier No. FN7913
This reusable feeding needle is designed for precise laboratory applications, featuring a curved 18-gauge stainless steel construction for durability and ease of use. It is engineered with a 2-inch length and a 2.25 mm ball diameter, providing reliable performance for animal research tasks.
  • Constructed from high-quality stainless steel for long-term reusability.
  • Features a curved design to facilitate targeted administration.
  • Equipped with a 2.25 mm ball tip for safe insertion.
  • Compatible with standard laboratory procedures requiring 18-gauge dimensions.