Learn More
Roboz Surgical REUSEABLE FEEDING NEEDLES
SDP
Supplier:  Roboz Surgical FN7910
Encompass
Reusable Feeding Needle; Stainless Steel; Curved; 20 Gauge; 1.5 in. Length; 2.25mm Ball Diameter
Catalog No. NC9178496