Learn More
Roboz Surgical REUSEABLE FEEDING NEEDLES
SDP
Catalog No. NC9178500
Encompass
Reusable Feeding Needle; Stainless Steel; Straight; 22 Gauge; 1.5 in. Length; 1.25mm Ball Diameter
Catalog No. NC9178500 Supplier Roboz Surgical Supplier No. FN7920
Reusable Feeding Needle; Stainless Steel; Straight; 22 Gauge; 1.5 in. Length; 1.25mm Ball Diameter