Learn More
ABclonal Technology Rev-Erba/NR1D1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A20452100UL
Encompass_Preferred
This gene encodes a transcription factor that is a member of the nuclear receptor subfamily 1 The encoded protein is a ligand-sensitive transcription factor that negatively regulates the expression of core clock proteins In particular this protein represses the circadian clock transcription factor aryl hydrocarbon receptor nuclear translocator-like protein 1 (ARNTL) This protein may also be involved in regulating genes that function in metabolic inflammatory and cardiovascular processes
Catalog No. 50-000-08358