Learn More
MSC RTS Chipboard Box, 4 x 4 x 8 in, 250/Pk.
Catalog No. 501915619
Encompass
These chipboard shipping boxes are designed for packaging small parts, featuring a reverse tuck folding carton design for positive locking. Constructed from .024 Kraft fibreboard, these boxes meet or exceed PPP-B-566 federal specifications.
  • Lightweight, ideal for small parts
  • Pye-lock tuck for secure closure
  • Die-cut from .024 fibreboard
  • Meets or exceeds PPP-B-566 federal specification
  • Made in USA
  • Single wall construction
Catalog No. 50-191-5619 Supplier MSC Supplier No. 39551189
These chipboard shipping boxes are designed for packaging small parts, featuring a reverse tuck folding carton design for positive locking. Constructed from .024 Kraft fibreboard, these boxes meet or exceed PPP-B-566 federal specifications.
  • Lightweight, ideal for small parts
  • Pye-lock tuck for secure closure
  • Die-cut from .024 fibreboard
  • Meets or exceeds PPP-B-566 federal specification
  • Made in USA
  • Single wall construction