Learn More
Medchemexpress LLC HY-100468 5mg Medchemexpress, REV7/REV3L-IN-1 CAS:1979192-13-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004685MG
Encompass_Preferred
Medchemexpress, HY-100468 5mg REV7/REV3L-IN-1 CAS:1979192-13-6 REV7/REV3L-IN-1 is a REV7/REV3L interaction inhibitor with an IC50 of 78 μM, which directly binds to REV7 in nuclear magnetic resonance analyses, and inhibits the reactivation of a reporter plasmid containing an interstrand crosslink (ICL) in between the promoter and reporter regions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0590