Learn More
Med Vet International Revalor S, 10 Implant Box
Catalog No. 502188475
Encompass_Preferred
Revalor-S is an implant containing 120mg of trenbolone acetate and 24mg of estradiol. Each implant consists of 6 small yellow pellets. Ten implants are provided in a cartridge. Increases rate of weight gain and improves feed efficiency in a slow-release delivery system.Products are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-218-8475 Supplier Med Vet International Supplier No. PHREVALORS
Revalor-S is an implant containing 120mg of trenbolone acetate and 24mg of estradiol. Each implant consists of 6 small yellow pellets. Ten implants are provided in a cartridge. Increases rate of weight gain and improves feed efficiency in a slow-release delivery system.Products are not always manufacturer specific. Actual product appearance may differ from website image.