Learn More
Medchemexpress LLC HY-N7067 10mg Medchemexpress, Revaprazan (hydrochloride) CAS:178307-42-1 Purity:>98%
SDP
Catalog No. 501970496
Encompass_Preferred
Medchemexpress, HY-N7067 10mg Revaprazan (HCl) CAS:178307-42-1 Revaprazan hydrochloride is a novel acid pump antagonist (APA). Revaprazan hydrochloride reduces COX-2 expression and has significant anti-inflammatory actions activities in H. pylori infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0496 Supplier Medchemexpress LLC Supplier No. HYN706710MG
Medchemexpress, HY-N7067 10mg Revaprazan (HCl) CAS:178307-42-1 Revaprazan hydrochloride is a novel acid pump antagonist (APA). Revaprazan hydrochloride reduces COX-2 expression and has significant anti-inflammatory actions activities in H. pylori infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.