Learn More
Medchemexpress LLC Revaprazan (hydrochloride) | 178307-42-1 | 1 ML
SDP
Catalog No. 5000418861
Encompass_Preferred
Revaprazan hydrochloride is a reversible proton pump inhibitor. It inhibits gastric acid secretion and protects gastric mucosa. It can also inhibit IkappaB-alpha degradation and Akt inactivation, leading to the attenuation of H. pylori-induced COX-2 expression. This product is used for research on infections and inflammation, such as H. pylori-infected gastric inflammation and gastric ulcers.
  • Reversible proton pump inhibitor
  • Inhibits gastric acid secretion
  • Protects gastric mucosa
  • Attenuates H. pylori-induced COX-2 expression
  • Reduces H. pylori-stimulated COX-2 expression and activity in AGS cells
  • Inhibits transcription activity of NF-κB in H. pylori-stimulated AGS cells
  • Improves retarded cell growth in H. pylori-stimulated AGS cells
  • Used for research on infection and inflammation
Catalog No. 50-004-18861 Supplier Medchemexpress LLC Supplier No. HYN706710MM/1ML
Revaprazan hydrochloride is a reversible proton pump inhibitor. It inhibits gastric acid secretion and protects gastric mucosa. It can also inhibit IkappaB-alpha degradation and Akt inactivation, leading to the attenuation of H. pylori-induced COX-2 expression. This product is used for research on infections and inflammation, such as H. pylori-infected gastric inflammation and gastric ulcers.
  • Reversible proton pump inhibitor
  • Inhibits gastric acid secretion
  • Protects gastric mucosa
  • Attenuates H. pylori-induced COX-2 expression
  • Reduces H. pylori-stimulated COX-2 expression and activity in AGS cells
  • Inhibits transcription activity of NF-κB in H. pylori-stimulated AGS cells
  • Improves retarded cell growth in H. pylori-stimulated AGS cells
  • Used for research on infection and inflammation