Learn More
Medchemexpress LLC Revaprazan hydrochloride | 178307-42-1 | 100 MG
SDP
Catalog No. 5000418956
Encompass_Preferred
Revaprazan hydrochloride is a reversible proton pump inhibitor that effectively inhibits gastric acid secretion and offers protection to the gastric mucosa. It functions by attenuating H. pylori-induced COX-2 expression through the inhibition of IκB-alpha degradation and Akt inactivation. This compound is suitable for research on infections and inflammation, including H. pylori-infected gastric inflammation and gastric ulcers.
  • Reversibly inhibits proton pumps, reducing gastric acid secretion.
  • Helps protect the stomach lining.
  • Attenuates H. pylori-induced COX-2 expression.
  • Achieves its effects by inhibiting IκB-alpha degradation and Akt inactivation.
  • Useful for studies on various infections and inflammation, such as H. pylori-infected gastric inflammation and gastric ulcers.
  • Available with a purity of 99.98%.
Catalog No. 50-004-18956 Supplier Medchemexpress LLC Supplier No. HYN7067100MG
Revaprazan hydrochloride is a reversible proton pump inhibitor that effectively inhibits gastric acid secretion and offers protection to the gastric mucosa. It functions by attenuating H. pylori-induced COX-2 expression through the inhibition of IκB-alpha degradation and Akt inactivation. This compound is suitable for research on infections and inflammation, including H. pylori-infected gastric inflammation and gastric ulcers.
  • Reversibly inhibits proton pumps, reducing gastric acid secretion.
  • Helps protect the stomach lining.
  • Attenuates H. pylori-induced COX-2 expression.
  • Achieves its effects by inhibiting IκB-alpha degradation and Akt inactivation.
  • Useful for studies on various infections and inflammation, such as H. pylori-infected gastric inflammation and gastric ulcers.
  • Available with a purity of 99.98%.