Learn More
Medchemexpress LLC Revaprazan (hydrochloride) | 178307-42-1 | 25 MG
SDP
Catalog No. 5000420356
Encompass_Preferred
Revaprazan hydrochloride is a reversible proton pump inhibitor that can inhibit gastric acid secretion and protect gastric mucosa. It can also inhibit IkappaB-alpha degradation and Akt inactivation, which attenuates H. pylori-induced COX-2 expression. This product can be used for research in infection and inflammation, including H. pylori-infected gastric inflammation and gastric ulcer.
  • Reduces H. pylori-stimulated COX-2 expression and activity in AGS cells (5-20 μM, 24 h).
  • Inhibits transcription activity of NF-κB in H. pylori-stimulated AGS cells (5-20 μM, 24 h).
  • Improves the retarded cell growth in H. pylori-stimulated AGS cells (5-20 μM, 24 h).
Catalog No. 50-004-20356 Supplier Medchemexpress LLC Supplier No. 25MGHYN7067
Revaprazan hydrochloride is a reversible proton pump inhibitor that can inhibit gastric acid secretion and protect gastric mucosa. It can also inhibit IkappaB-alpha degradation and Akt inactivation, which attenuates H. pylori-induced COX-2 expression. This product can be used for research in infection and inflammation, including H. pylori-infected gastric inflammation and gastric ulcer.
  • Reduces H. pylori-stimulated COX-2 expression and activity in AGS cells (5-20 μM, 24 h).
  • Inhibits transcription activity of NF-κB in H. pylori-stimulated AGS cells (5-20 μM, 24 h).
  • Improves the retarded cell growth in H. pylori-stimulated AGS cells (5-20 μM, 24 h).