Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ Elkskin Leather Welder's Gloves with Nomex™ Lining

Catalog No. 19840317
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-840-317 X-Large
19-840-315 Medium
19-840-316 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-840-317 Supplier Black Stallion™ Supplier No. 850XL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extra thick, elkskin leather construction provides heat, flame, and abrasion resistance. DuPont™ Nomex™ liner on the back of the glove offers additional fire protection.

  • Elkskin leather construction is extra thick to provide heat, flame, and abrasion resistance
  • DuPont™ Nomex liner offers additional fire-resistant protection
  • Aramid Fiber stitching for spark and abrasion resistance
  • Reversed grain leather palm enhances grip, and provides a softer feel on the hand
  • Thumb strap provides additional reinforcement or high stress areas

Specifications

Color Gold
Material Elkskin Leather
Size X-Large
For Use With (Application) Stick Welding
Glove Cuff Color White
Glove Cuff Style Gauntlet
Liner Material Nomex™
Palm Type Grain Leather
Series 850
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.