Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Revco™ FMS High-Performance Lab Refrigerators
GSA_VA

Catalog No. RFMS2305A Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
651 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
RFMS2305A 651 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. RFMS2305A Supplier Thermo Scientific™ Supplier No. RFMS2305A

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Protect flammable materials, solvents and intermediates using Revco™ FMS High-Performance Laboratory Refrigerator.

Protect flammable materials, solvents and intermediates using Thermo Scientific™ Revco FMS High-Performance Laboratory Refrigerators. These high-performance refrigerators are designed to meet stringent US Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids.† These refrigerators are designed to maintain a safe environment while keeping valuable samples within a tight temperature range. Each is listed by UL as a special purpose refrigerator intended for flammable material storage in accordance with NFPA 45, 70 and 99.† 45, 70 and 99.†

  • Easy to use intuitive digital display featuring microprocessor control
  • Lockable doors, audio and visual alarms including door ajar alarms for security and peace of mind
  • Convenient access port and casters
  • Outstanding construction featuring painted, high-quality steel exteriors with durable finish
  • Ignition-free interior cabinet design for the safe storage of flammable materials
  • Meets safety standards in accordance with UL, NFPA† and OSHA†for regulatory compliance
  • Optional, built-in, inkless, seven-day graphic chart recorder
  • Optional, barrier-protected 4-20 milliamp output for external monitoring
  • Manual defrost

Ordering Information

Temperature chart recorders, and 4-20 milliamp are available separately. Not Available Outside North America

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Flammable Storage Refrigerator
Capacity (English) 23 cu. ft.
Capacity (Metric) 651 L
Temperature Range 1°C to 8°C
Door Style Solid
No. of Doors 1
Shelves 4
Cabinet Material Spark-free Steel
Exterior Finish Painted
Certifications/Compliance UL
Style Built-in
Defrost Manual
Monitoring Options Alarm, Recorder
Static Holding Time 7 Days
Compressor Hermetic
Depth (English) Interior 29 in.
Depth (Metric) Interior 74 cm
Height (English) Exterior 78.6 in.
Height (English) Interior 58 in.
Height (Metric) Exterior 200 cm
Height (Metric) Interior 147 cm
Length (English) Exterior 37 in.
Length (Metric) Exterior 94 cm
Width (English) Exterior 28 in.
Width (English) Interior 24 in.
Width (Metric) Exterior 72 cm
Width (Metric) Interior 61 cm
Amperage 4A
Breaker 15A
Plug Type NEMA 5-15
Voltage 115 V
Frequency 60 Hz
Shipping Weight (English) 425 lb.
Shipping Weight (Metric) 193 kg
Show More Show Less

Note:

† Products complying with the applicable requirements of NFPA 70, National Electric Code, NPFA 45, Fire Protection for Laboratories Using Chemicals, and NFPA 99 Health Care Facilities are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.