Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ 22 oz. PBI High Temperature Gloves - 23 in.

Catalog No. 19056496
Encompass_Preferred
Change view
Click to view available options
Hand:
Specific
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hand
19-056496 Specific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-056496 Supplier Black Stallion™ Supplier No. P123
This item is not returnable. View return policy

Gloves are constructed with 22 oz. PBI and feature wool insulation for enhanced protection during high heat applications.

  • Constructed of 22 oz. PBI with wool lining
  • 23 in. cuff extends protection past the wrist for additional thermal protection

Specifications

Color Natural
Hand Specific
Material PBI
Glove Cuff Style Open
Liner Material Wool
Series 123
Yarn Weight 22 oz.
Length (English) 23 in.
Length (Metric) 58.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.