Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ Side Split Cowhide Leather Welder's Gloves with CushionCore™ Liner

Catalog No. 19001503
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-001-503 Medium
19-001-506 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-001-503 Supplier Black Stallion™ Supplier No. 320M
Add to Cart
Add to Cart
This item is not returnable. View return policy

CushionCore™ padded liner provides heat-blocking comfort, while Aramid Fiber stitching protects against sparks and abrasion.

  • Side split cowhide leather construction is durable and flexible
  • CushionCore™ padded liner provides heat-blocking comfort
  • Aramid Fiber stitching for spark and abrasion resistance
  • Wide straps on palm for additional reinforcement in a key stress area
  • Welted seams ensure maximum protection for threads

Specifications

Color Blue, Gray
Hand Specific
Material Side Split Cowhide Leather
Size Medium
For Use With (Application) Stick Welding
Glove Cuff Style Gauntlet
Series 320
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.