Learn More
NEOGEN Reveal 2.0 for E. coli O157:H7, Complete System
SDP
Supplier:  NEOGEN 700002798
Encompass_Preferred
The Reveal 2.0 for E. coli O157:H7 test provides for the rapid recovery of E. coli O157:H7 and E. coli O157:NM organisms in beef matrices facilitating detection and presumptive identification of the target organisms in as little as 12 hours following enrichment.Approvals: AOAC-RI 011103Limit of Detection: 1 cfu/analytical unitTesting Time: 10 MinutesFor more information visit https://www.neogen.com/solutions/microbiology/reveal-2-e-coli-o157-h7
Catalog No. 50-201-5125