Learn More
NEOGEN Reveal 2.0 for Listeria, One-Step Complete System for Foods
SDP
Supplier:  NEOGEN 700002829
Encompass_Preferred
The Reveal 2.0 for Listeria test provides for the rapid recovery of Listeria in food and environmental samples allowing next day detection and presumptive identification of Listeria.Approvals: AOAC-RI 041101Limit of Detection: 1 cfu/analytical unitTesting Time: 20 MinutesTime to Result: 27-30 HoursFor more information visit https://www.neogen.com/solutions/microbiology/reveal-2-listeria/
Catalog No. 50-201-5135