Learn More
NEOGEN Reveal 3-D for Almond
SDP
Supplier:  NEOGEN 700002645
Encompass_Preferred
Reveal 3-D for Almond offers results in just 5 minutes and can be used in virtually any environment to screen environmental swabs and clean-in-place rinses. The Reveal 3-D line of food allergen tests kits is uniquely designed to guard against oversaturation and false negative results, with results displayed in three visual lines on the lateral flow test strip. A control line assures the test is working properly, and two other lines outline nondetectable, low, and high levels of almond.For more information visit https://www.neogen.com/solutions/Allergens/reveal-3d-almond
Catalog No. 50-201-5150