Learn More
NEOGEN Reveal 3-D for Crustacea
Supplier: NEOGEN 700002641
Reveal 3-D for Crustacea offers results in just 5 minutes and can be used in virtually any environment to screen environmental swabs and clean-in-place rinses. The Reveal 3-D line of food allergen tests kits is uniquely designed to guard against over-saturation and false negative results, with results displayed in three visual lines on the lateral flow test strip. A control line assures the test is working properly, and two other lines outline non-detectable, low, and high levels of crustacea.For more information visit https://www.neogen.com/solutions/Allergens/reveal-3d-crustacea