Learn More
NEOGEN Reveal 3-D for Total Milk
SDP
Supplier:  NEOGEN 700002580
Encompass_Preferred
Reveal 3-D for Total Milk offers results in just 5 minutes and can be used in virtually any environment to screen for milk residue (casein or whey) with environmental swabs, liquid products (such as juices and sorbet) and clean-in-place rinses. The Reveal 3-D line of food allergen tests kits is uniquely designed to guard against oversaturation and false negative results, with results displayed in three visual lines on the lateral flow test strip. A control line assures the test is working properly, and two other lines outline nondetectable, low, and high levels of total milk.For more information visit https://www.neogen.com/solutions/Allergens/reveal-3d-total-milk
Catalog No. 50-201-5117