Learn More
Description
- The electronic low airflow monitoring system includes a manual speed controller, hour counter, and electronic blower switch.
- The Multiplex™ Filtration System includes a leakage prevention clamp, low airflow alarm, filter door key, and ID window.
- 1 FS027 HEPA filter
- 1 FS028 Electrostatic Pre-filter
- Ebmpapst™ centrifugal fan
- 270° visibility
- Utilities or Power: Available in two power options:120 V/60 Hz and 220 V/50 Hz
- Constructed with Epoxy-coated steel, a perforated stainless steel surface,Vapor-proof interior fluorescent lighting, and a polypropylene Spill-retention tray.
- Options Accessories
- FS027: HEPA filter
- FS028: Electrostatic Pre-filters
- Compatible with the FS029 Mobile Cart.
- Pulls hazardous vapors into the station and away from the user without releasing them into the environment
- Keeps fumes separate from internal cabinet components
- Provides unrestricted access to the front and sides of the work area
- Provides focused lighting
- Noise level: Less than 52 dBA at 1 meter
- Airflow: 145 CFM
- Face velocity: 80 FPM
- Uses downward airflow and advanced filtration to help protect the user.
- Maintains a clear, Open-access work area.
- Supports efficient evidence processing and laboratory work.
Specifications
Specifications
| Product Type | Ductless Downflow Workstation |
| Filter Type | Prefilter, HEPA |
| Material | Polypropylene |
| Dimensions (D x W x H) | 22.42 x 24 x 35.5 in. |
| Exhaust | Ductless |
| Height (English) Interior | 15.75 in. |
| Includes | 1 Electrostatic Pre Filter and 1 HEPA Filter |
| Voltage | 120/220 V |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
